missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals

Catalog Number 10397820
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
32503-27-8
C16H37NO4S
339.54
MFCD00011637
SHFJWMWCIHQNCP-UHFFFAOYSA-M
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
94433
hydrogen sulfate;tetrabutylazanium
OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
32503-27-8
C16H37NO4S
339.54
MFCD00011637
SHFJWMWCIHQNCP-UHFFFAOYSA-M
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
94433
hydrogen sulfate;tetrabutylazanium
OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 168.0°C to 172.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula [CH3(CH2)3]4NHSO4
Quantity 1 kg
Fieser 06,565; 07,354; 13,94
Solubility Information Solubility in water: >50%.
Formula Weight 339.53
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Tetrabutylammonium hydrogen sulfate, 98%
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'