missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hexafluorophosphate, TRC

Product Code. 30452559
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452559 10 g -
30450480 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452559 Supplier Toronto Research Chemicals Supplier No. TRCT31065810G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3109-63-5
Molecular Formula C16H36F6NP
Molecular Weight (g/mol) 387.43
IUPAC Name tetrabutylazanium;hexafluorophosphate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.F[P-](F)(F)(F)(F)F

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H36N.F6P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-7(2,3,4,5)6/h5-16H2,1-4H3;/q+1;-1
Formula Weight 387.249
Chemical Name or Material Tetrabutylammonium hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.