missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Chloride, TRC

Product Code. 30428426
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428426 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428426 Supplier Toronto Research Chemicals Supplier No. TRCT29122525G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium Chloride
CAS 1112-67-0
Molecular Formula C16H36ClN
Quantity 25 g
Synonym N,N,N-Tributyl-1-butanaminium Chloride, TBAC, TBAC 100, Tetra-n-butylammonium Chloride
InChI Formula InChI=1S/C16H36N.ClH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;chloride
Molecular Weight (g/mol) 277.92
Formula Weight 277.2536
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.