missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium chloride, 95%, Tech., contains also bromide

Product Code. 10792924 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10792924 500 g Glass bottle 500g
10532391 5 g Glass bottle 5g
10624852 25 g Glass bottle 25g
10614662 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10792924 Supplier Thermo Scientific Acros Supplier No. 174125000
  • ONLINE EXCLUSIVE PRICE
    £865.00
    £835.00 / 500g
    Save £30.00
    3% Off
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1112-67-0 | C16H36ClN | 277.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-67-0
Molecular Formula C16H36ClN
Molecular Weight (g/mol) 277.92
MDL Number MFCD00011635
InChI Key NHGXDBSUJJNIRV-UHFFFAOYSA-M
Synonym tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
PubChem CID 70681
ChEBI CHEBI:51988
IUPAC Name tetrabutylazanium;chloride
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Cl-]

Specifications

Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]4NCl
Quantity 500 g
Beilstein 04, III, 292
Fieser 06,565; 07,353; 17,363
Solubility Information Solubility in water: soluble.
Formula Weight 277.92
Percent Purity 95%
Grade Technical
Physical Form Crystalline Solid
Chemical Name or Material Tetrabutylammonium chloride, Also contains bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.