missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Bromide, TRC

Product Code. 30424518
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424518 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424518 Supplier Toronto Research Chemicals Supplier No. TRCT291315250G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium Bromide
CAS 1643-19-2
Purity Grade Notes HPLC
Molecular Formula C16 H36 N . Br
Quantity 250 g
Synonym 1-Butanaminium','N,N,N-tributyl-','bromide (1:1), 1-Butanaminium','N,N,N-tributyl-','bromide (9CI), Ammonium','tetrabutyl-','bromide (8CI), Tetrabutylammonium bromide (6CI,7CI), AP 6G, Actiron 43-65, Aliquat 100, Mastermix MB 4988, TBAB, TBAB 50A, Tetra-n-butylammonium bromide
InChI Formula InChI=1S/C16H36N.BrH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium;bromide
Molecular Weight (g/mol) 322.37
Formula Weight 321.2031
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.