missing translation for 'onlineSavingsMsg'
Learn More

Sodium hypochlorite, 4-6%

Product Code. 10667381 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
25mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10667381 25 mL Plastic bottle 25mL
10573094 250 mL Plastic bottle 250mL
10527752 1 L Plastic bottle 1 L
11974211 2.5 L Plastic bottle 2.5 L
15274734 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10667381 Supplier Thermo Scientific Acros Supplier No. 419550250
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 7681-52-9 | ClNaO | 74.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Color Yellow
Physical Form Solution
Quantity 25 mL
Linear Formula NaOCl
Fieser 01,1084; 04,456; 05,617; 06,543; 07,337; 08,461; 09,430; 10,365; 11,487; 15,293; 16,308
Merck Index 15,8762
Solubility Information Solubility in water: soluble
Formula Weight 74.44
Percent Purity 4.5 to 5.5%
Grade Reagent
Packaging Plastic bottle
Specific Gravity 1.083
Density 1.0830g/mL
Chemical Name or Material Sodium hypochlorite, 4-6% Chlorine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium hypochlorite, 5% active chlorine
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Sodium hypochlorite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.