missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Borohydride, TRC

Product Code. 30424517
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424517 25 g
30419814 5 g
30409459 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424517 Supplier Toronto Research Chemicals Supplier No. TRCT28923525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33725-74-5
Molecular Formula C16H40BN
Molecular Weight (g/mol) 257.3
Synonym tetrahydro-Borate(1-) N,N,N-tributyl-1-butanaminium, Tetrabutylammonium Borohydride, Tetrabutylammonium Tetrahydroborate, Tetrabutylammonium tetrahydroborate(1-)
IUPAC Name boranuide;tetrabutylazanium
SMILES [BH4-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C16H36N.BH4/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H4/q+1;-1
Formula Weight 257.3254
Chemical Name or Material Tetrabutylammonium Borohydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.