missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutyl Ammonium Bromide, for HPLC, Certified, Fisher Chemical

Product Code. 10214500 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10214500 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10214500 Supplier Fisher Chemical Supplier No. T/0476/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1643-19-2
Melting Point 100°C
Color Cream
Packaging Plastic powder jar
Molecular Formula C16H36BrN
MDL Number 11633
Quantity 25 g
Synonym tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r
InChI Key JRMUNVKIHCOMHV-UHFFFAOYSA-M
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Br-]
IUPAC Name tetrabutylazanium;bromide
Molecular Weight (g/mol) 322.375
PubChem CID 74236
ChEBI CHEBI:51993
Formula Weight 322.36g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.