missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromothiophene, 99+%, ACROS Organics™

Product Code. 10021620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10021620 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10021620 Supplier Thermo Scientific Acros Supplier No. 137960250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3958-03-0
Melting Point 112.0°C to 116.0°C
Color Brown to Cream
Boiling Point 326.0°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C4Br4S
MDL Number MFCD00005419
Quantity 25 g
Beilstein 17, 34
Synonym tetrabromothiophene, perbromothiophene, thiophene, tetrabromo, tetrabromthiophen, 2,3,4,5-tetrabromo-thiophene, pubchem7661, acmc-1ae1j, 2,4,5-tetrabromothiophene, ksc491i3p, 2,3,4,5-tetrabromothiophen
InChI Key AVPWUAFYDNQGNZ-UHFFFAOYSA-N
SMILES C1(=C(SC(=C1Br)Br)Br)Br
IUPAC Name 2,3,4,5-tetrabromothiophene
Molecular Weight (g/mol) 399.72
PubChem CID 77565
Formula Weight 399.71
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabromothiophene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabromothiophene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.