missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophthalic Anhydride, TRC

Product Code. 30427698
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427698 5 g 5g
30420781 100 g -
30410762 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427698 Supplier Toronto Research Chemicals Supplier No. TRCT2911805G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-79-1
Molecular Formula C8 Br4 O3
Molecular Weight (g/mol) 463.6998
Synonym Phthalic anhydride, tetrabromo- (6CI,7CI,8CI), 4,5,6,7-Tetrabromo-1,3-isobenzofurandione, 3,4,5,6-Tetrabromophthalic anhydride, 4,5,6,7-Tetrabromoisobenzofuran-1,3-dione, Bromphthal, FG 4000, FireMaster PHT 4, NSC 4874, PHT 4, RB 49, Saytex RB 49, Tertrabromophthalic anhydride, Tetrabromophthalic acid anhydride, Tetrabromophthalic anhydride
IUPAC Name 4,5,6,7-tetrabromo-2-benzofuran-1,3-dione
SMILES Brc1c(Br)c(Br)c2C(=O)OC(=O)c2c1Br

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8Br4O3/c9-3-1-2(8(14)15-7(1)13)4(10)6(12)5(3)11
Formula Weight 459.6581
Chemical Name or Material Tetrabromophthalic anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.