missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophthalic Anhydride 98%, Thermo Scientific™

Catalog Number 10407150
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 632-79-1
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C8Br4O3
MDL Number MFCD00005919
Quantity 5g
Synonym tetrabromophthalic anhydride, bromphthal, firemaster pht 4, phthalic anhydride, tetrabromo, bromophthal, tetrabromophthalic acid anhydride, firemaster pht4, 1,3-isobenzofurandione, 4,5,6,7-tetrabromo, 4,5,6,7-tetrabromo-1,3-isobenzofurandione, pht 4
Solubility Information Solubility in water: insoluble (hydrolysis to acid). Other solubilities: insoluble in ethanol,, slightly soluble in benzene,, soluble in xylene, acetic acid
InChI Key QHWKHLYUUZGSCW-UHFFFAOYSA-N
SMILES C12=C(C(=C(C(=C1Br)Br)Br)Br)C(=O)OC2=O
IUPAC Name 4,5,6,7-tetrabromo-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 463.701
PubChem CID 12443
Formula Weight 463.7
Percent Purity 98%
Chemical Name or Material Tetrabromophthalic anhydride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabromophthalic anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'