missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophenolphthalein Ethyl Ester, Potassium Salt, Indicator Grade, Thermo Scientific Chemicals

Product Code. 10216200
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-91-6
Molecular Formula C22H13Br4KO4
Molecular Weight (g/mol) 700.06
MDL Number MFCD00011662
InChI Key WCIQBKUTYDIBJC-UHFFFAOYSA-M
PubChem CID 23689366
SMILES [K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1

Specifications

Merck Index 15, 9331
Infrared Spectrum Authentic
IUPAC Name potassium;2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)-(2-ethoxycarbonylphenyl)methyl]phenolate
Formula Weight 700.05
Grade Extra Pure
Packaging Glass bottle
Specific Extinction 660 min. (600nm, ethanol)
Color Black/Blue to Blue
Quantity 1 g
Physical Form Crystalline Powder or Granules
Chemical Name or Material Tetrabromophenolphthalein ethyl ester, potassium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'