missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromocatechol, 96%

Product Code. 11300174
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11300174

Brand: Thermo Scientific Alfa Aesar L00690.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 488-47-1 | C6H2Br4O2 | 425.70 g/mol

Tetrabromocatechol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromocatechol is used as a pharmaceutical intermediate.

Solubility
It is partly miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong bases. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-47-1
Molecular Formula C6H2Br4O2
Molecular Weight (g/mol) 425.70
MDL Number MFCD00002189
InChI Key OAUWOBSDSJNJQP-UHFFFAOYSA-N
Synonym tetrabromocatechol, tetrabromopyrocatechol, pyrocatechol, tetrabromo, 1,2-benzenediol, 3,4,5,6-tetrabromo, 3,4,5,6-tetrabromocatechol, unii-n642cph23m, 1, 3,4,5,6-tetrabromo, 4-06-00-05624 beilstein handbook reference
PubChem CID 61127
IUPAC Name 3,4,5,6-tetrabromobenzene-1,2-diol
SMILES OC1=C(O)C(Br)=C(Br)C(Br)=C1Br

Specifications

Melting Point 189°C to 193°C
Quantity 5 g
Beilstein 2051548
Solubility Information It is partly miscible with water.
Formula Weight 425.7
Percent Purity 96%
Chemical Name or Material Tetrabromocatechol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.