missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromobisphenol A Allyl Ether, TRC

Product Code. 30424512
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30424512 500 g
30409201 10 g
30418633 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424512 Supplier Toronto Research Chemicals Supplier No. TRCT291095500G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25327-89-3
Molecular Formula C21 H20 Br4 O2
Molecular Weight (g/mol) 624
Synonym Benzene, 1,1'-(1-methylethylidene)bis[3,5-dibromo-4-(2-propenyloxy)- (9CI), Propane, 2,2-bis[4-(allyloxy)-3,5-dibromophenyl]- (8CI), 1,1'-(1-Methylethylidene)bis[3,5-dibromo-4-(2-propen-1-yloxy)benzene], 1,1'-Isopropylidenebis[4-(allyloxy)-3,5-dibromobenzene], 2,2-Bis(3,5-dibromo-4-allyloxyphenyl)propane, 2,2-Bis(4-allyloxy-3,5-dibromophenyl)propane, BE 51, FG 3200, Fire Guard 3200, Flame Cut 122K, Pyroguard SR 319, SR 319, TBBPA-DE, Tetrabromobisphenol A allyl ether, Tetrabromobisphenol A bis(allyl ether), Tetrabromobisphenol A diallyl ether
IUPAC Name 1,3-dibromo-5-[2-(3,5-dibromo-4-prop-2-enoxyphenyl)propan-2-yl]-2-prop-2-enoxybenzene
SMILES CC(C)(c1cc(Br)c(OCC=C)c(Br)c1)c2cc(Br)c(OCC=C)c(Br)c2

Specifications

Quantity 500 g
InChI Formula InChI=1S/C21H20Br4O2/c1-5-7-26-19-15(22)9-13(10-16(19)23)21(3,4)14-11-17(24)20(18(25)12-14)27-8-6-2/h5-6,9-12H,1-2,7-8H2,3-4H3
Formula Weight 619.8197
Chemical Name or Material Tetrabromobisphenol A diallyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.