missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenzyl Pyrophosphate, TRC

Product Code. 30428677
Change view
Click to view available options
Quantity:
1 g
2.5 g
10 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428677 2.5 g 2.5g
30412593 1 g -
30424115 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428677 Supplier Toronto Research Chemicals Supplier No. TRCT2894502.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 990-91-0
Molecular Formula C28 H28 O7 P2
Molecular Weight (g/mol) 538.47
Synonym Diphosphoric Acid P,P,P',P'-Tetrakis(phenylmethyl) Ester, Benzyl Pyrophosphate, Tetrabenzyl Diphosphate
IUPAC Name dibenzyl bis(phenylmethoxy)phosphoryl phosphate
SMILES O=P(OCc1ccccc1)(OCc2ccccc2)OP(=O)(OCc3ccccc3)OCc4ccccc4

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C28H28O7P2/c29-36(31-21-25-13-5-1-6-14-25,32-22-26-15-7-2-8-16-26)35-37(30,33-23-27-17-9-3-10-18-27)34-24-28-19-11-4-12-20-28/h1-20H,21-24H2
Formula Weight 538.131
Chemical Name or Material Tetrabenzyl Pyrophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.