missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenazine Racemate, MedChemExpress

Product Code. 16498376
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498376 100 mg 100mg
16478538 10 mg 10mg
16454407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498376 Supplier Medchem Express Supplier No. HYB0590A
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tetrabenazine Racemate (Ro 1-9569 Racemate) is a selective and reversible inhibitor of vesicular monoamine transporter-2 (VMAT-2).

Tetrabenazine Racemate (Ro 1-9569 Racemate) is a selective and reversible inhibitor of vesicular monoamine transporter-2 (VMAT-2).

Specifications

Chemical Name or Material Tetrabenazine Racemate
CAS 718635-93-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H27NO3
Quantity 100 mg
Synonym Ro 1-9569 Racemate
Solubility Information DMSO : 50 mg/mL (157.52 mM; Need ultrasonic)
SMILES O=C1C(CC(C)C)CN2CCC3=CC(OC)=C(OC)C=C3C2C1
Molecular Weight (g/mol) 317.42
Formula Weight 317.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.