missing translation for 'onlineSavingsMsg'
Learn More

(+)-Tetrabenazine, MedChemExpress

Product Code. 16488538
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488538 10 mg 10mg
16464407 50 mg 50mg
16431018 5 mg 5mg
16408386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488538 Supplier Medchem Express Supplier No. HYB0590B
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress (+)-Tetrabenazine ((+)-TBZ; (3R,11bR)-TBZ; (3R,11bR)-Tetrabenazine) is a reversible inhibitor of vesicular monoamine transporter 2 (VMAT-2), inhibits transport by VMAT2 with 10-fold greater potency than transport by VMAT1.

(+)-Tetrabenazine ((+)-TBZ

Specifications

Chemical Name or Material (+)-Tetrabenazine
CAS 1026016-83-0
Color White
Purity Grade Notes Research
Molecular Formula C19H27NO3
Quantity 10 mg
Synonym (+)-TBZ (3R,11bR)-TBZ (3R,11bR)-Tetrabenazine
Solubility Information DMSO : 16.67 mg/mL (52.52 mM; Need ultrasonic)
SMILES COC1=C(OC)C=C(CCN(C[C@H]2CC(C)C)[C@]3([H])CC2=O)C3=C1
Molecular Weight (g/mol) 317.42
Formula Weight 317.42
Percent Purity 95.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.