missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenazine-d6, MedChemExpress

Product Code. 16498538
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498538 10 mg 10mg
16482239 1 mg 1mg
16441018 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498538 Supplier Medchem Express Supplier No. HYB0590S
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tetrabenazine D6 is the deuterium labeled Tetrabenazine, which is a VMAT-inhibitor used for treatment of hyperkinetic movement disorder.

Tetrabenazine D6 is the deuterium labeled Tetrabenazine, which is a VMAT-inhibitor used for treatment of hyperkinetic movement disorder.

Specifications

Chemical Name or Material Tetrabenazine-d6
CAS 1392826-25-3
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C19H21D6NO3
Color Off-White
Synonym Ro 1-9569-d6
SMILES O=C1[C@H](CC(C)C)CN2CCC3=CC(OC([2H])([2H])[2H])=C(OC([2H])([2H])[2H])C=C3[C@@]2([H])C1
Industry Type Neuroscience-Neuromodulation
Molecular Weight (g/mol) 323.46
Quantity 10 mg
Formula Weight 323.46
Percent Purity 98.3%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.