missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) nitrate solution, Pt 3-4% w/w (cont. Pt)

Product Code. 11335919
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335919 2 g 2g
11345919 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335919 Supplier Thermo Scientific Alfa Aesar Supplier No. 012167.NA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20634-12-2 | H12N6O6Pt | 387.22 g/mol

Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20634-12-2
Molecular Formula H12N6O6Pt
Molecular Weight (g/mol) 387.22
MDL Number MFCD00011624
InChI Key RBAKORNXYLGSJB-UHFFFAOYSA-N
Synonym tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt
PubChem CID 146543
IUPAC Name azane;platinum(2+);dinitrate
SMILES N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 2 g
Linear Formula (NH3)4Pt(NO3)2
UN Number UN3218
Solubility Information Miscible with water.
Formula Weight 387.21
Concentration or Composition (by Analyte or Components) Pt 3 to 4% w/w (cont. Pt)
Density 1.05g/mL
Chemical Name or Material Tetraammineplatinum(II) nitrate solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.