missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) nitrate

Product Code. 11319359
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11319359

Brand: Thermo Scientific Alfa Aesar 088960.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20634-12-2 | H12N6O6Pt | 387.22 g/mol

Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20634-12-2
Molecular Formula H12N6O6Pt
Molecular Weight (g/mol) 387.22
MDL Number MFCD00011624
InChI Key RBAKORNXYLGSJB-UHFFFAOYSA-N
Synonym tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt
PubChem CID 146543
IUPAC Name azane;platinum(2+);dinitrate
SMILES N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 262°C (decomposition)
Physical Form Chunks and/or Powder
Quantity 5 g
Solubility Information Miscible with water.
Formula Weight 387.21
Chemical Name or Material Tetraammineplatinum(II) nitrate
compliance-icons compliance-icons
Product Identifier
  • Tetraammineplatinum(II) nitrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Tetraammineplatinum(II) nitrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.