missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) hydroxide solution, Pt 8-11% w/w (cont. Pt)

Product Code. 11336169
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336169 1 Ea. 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11336169 Supplier Thermo Scientific Alfa Aesar Supplier No. 042918.ND
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38201-97-7 | H{1}{4}N{4}O{2}Pt

Tetraammineplatinum(II) hydroxide solution acts as a precursor for a platinum catalyst. It acts as noble metal catalyst which is primarily useful for homogeneous catalysis applications. It is used as a catalyst in hydrogenation, carbonylation, hydroformylation, chiral catalysis reactions and ruthenium dyes. Furthermore, it is used for C-X coupling reaction like Heck, Suzuki and Stille reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38201-97-7
Physical Form Liquid
Quantity 1 Ea.
Molecular Formula H{1}{4}N{4}O{2}Pt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.