missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) hydrogen phosphate solution, Pt 2% w/w (cont. Pt), Thermo Scientific™

Product Code. 11460660
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460660 5 g 5g
11470660 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460660 Supplier Thermo Scientific Alfa Aesar Supplier No. 044047.NB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127733-98-6
Molecular Formula H13N4O4PPt
Molecular Weight (g/mol) 359.19
MDL Number MFCD03788258
InChI Key LSDWMQXWIQMHNQ-UHFFFAOYSA-L
Synonym azane;hydrogen phosphate;platinum 2+, platinum 2+ tetraamine hydrogen phosphate, platinum 2+ hydrogen phosphate-ammonia 1/1/4, platinum 2+ ion tetraamine hydrogen phosphate
IUPAC Name platinum(2+) tetraamine hydrogen phosphate
SMILES N.N.N.N.[Pt++].OP([O-])([O-])=O

Specifications

Color Colorless
Quantity 5 g
Solubility Information Fully miscible with water.
Formula Weight 359.18 (Anhydrous)
Concentration or Composition (by Analyte or Components) Pt 2% w/w (cont. Pt)
Physical Form Liquid
Chemical Name or Material Tetraammineplatinum (II) hydrogen phosphate solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.