missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) hydrogen carbonate, Thermo Scientific Chemicals

Product Code. 15411957
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123439-82-7
Molecular Formula C2H16N4O6Pt
Molecular Weight (g/mol) 387.26
MDL Number MFCD03788256
InChI Key AGUMVCVDCJQCIQ-UHFFFAOYSA-N
Synonym tetraammineplatinum ii hydrogen carbonate, platinum tetrammine hydrogencarbonate, c2h14n4o6pt, tetraammineplatinum ii hydrogencarbonate, carboxyoxy platinio hydrogen carbonate tetraamine, tetraammineplatinum ii hydrogencarbonate trace metals basis
IUPAC Name bis(carbonic acid) tetraamine platinum
SMILES N.N.N.N.[Pt].OC(O)=O.OC(O)=O

Specifications

Melting Point 236°C (decomposition)
Color White
Density 2.7 g/mL at 4°C
Linear Formula [Pt(NH3)4](HCO3)2
Quantity 1 g
Formula Weight 385.24
Physical Form Crystalline
Chemical Name or Material Tetraammineplatinum (II) hydrogen carbonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'