missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) hydrogen carbonate, Thermo Scientific Chemicals

£152.00 - £446.00

Chemical Identifiers

CAS 123439-82-7
Molecular Formula C2H16N4O6Pt
Molecular Weight (g/mol) 387.26
MDL Number MFCD03788256
InChI Key AGUMVCVDCJQCIQ-UHFFFAOYSA-N
Synonym tetraammineplatinum ii hydrogen carbonate, platinum tetrammine hydrogencarbonate, c2h14n4o6pt, tetraammineplatinum ii hydrogencarbonate, carboxyoxy platinio hydrogen carbonate tetraamine, tetraammineplatinum ii hydrogencarbonate trace metals basis
IUPAC Name bis(carbonic acid) tetraamine platinum
SMILES N.N.N.N.[Pt].OC(O)=O.OC(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15411957
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044042.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15421957
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044042.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123439-82-7
387.26
AGUMVCVDCJQCIQ-UHFFFAOYSA-N
bis(carbonic acid) tetraamine platinum
C2H16N4O6Pt
MFCD03788256
tetraammineplatinum ii hydrogen carbonate, platinum tetrammine hydrogencarbonate, c2h14n4o6pt, tetraammineplatinum ii hydrogencarbonate, carboxyoxy platinio hydrogen carbonate tetraamine, tetraammineplatinum ii hydrogencarbonate trace metals basis
N.N.N.N.[Pt].OC(O)=O.OC(O)=O

Specifications

123439-82-7
White
C2H16N4O6Pt
MFCD03788256
AGUMVCVDCJQCIQ-UHFFFAOYSA-N
bis(carbonic acid) tetraamine platinum
385.24
Tetraammineplatinum (II) hydrogen carbonate
236°C (decomposition)
2.7 g/mL at 4°C
[Pt(NH3)4](HCO3)2
tetraammineplatinum ii hydrogen carbonate, platinum tetrammine hydrogencarbonate, c2h14n4o6pt, tetraammineplatinum ii hydrogencarbonate, carboxyoxy platinio hydrogen carbonate tetraamine, tetraammineplatinum ii hydrogencarbonate trace metals basis
N.N.N.N.[Pt].OC(O)=O.OC(O)=O
387.26
Crystalline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

missing translation for 'einecsNumber' : 426-730-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only