missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) chloride monohydrate

Product Code. p-7048441
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11336058

Brand: Thermo Scientific Alfa Aesar 010544.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13933-33-0 | Cl2H14N4OPt | 352.12 g/mol

Employed in the synthesis of cisplatin, and platinum included carbon - carbon materials with metal/metal oxide nanoparticles. Useful for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13933-33-0
Molecular Formula Cl2H14N4OPt
Molecular Weight (g/mol) 352.12
MDL Number MFCD00149947
InChI Key GWBDNMCYHWRSOH-UHFFFAOYSA-L
Synonym azane;platinum 2+ ;dichloride;hydrate, tetrammineplatinum ii chloride hydrate, tetraammineplatinum ii chloride monohydrate, platinum 2+ ion tetraamine hydrate dichloride, platinum 2+ tetraamine hydrate dichloride, platinum 2+ chloride-ammonia-water 1/2/4/1, tetraamineplatinium ii chloride
PubChem CID 21946612
IUPAC Name azane;platinum(2+);dichloride;hydrate
SMILES N.N.N.N.O.[Cl-].[Cl-].[Pt++]

Specifications

Melting Point 250°C (dec.)
Color White through Cream
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water,ethanol and chloroform.
Formula Weight 352.13 (334.11 Anhydrous)
Sensitivity Hygroscopic
Density 2.74g/mL
Chemical Name or Material Tetraammineplatinum(II) chloride monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.