missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) chloride monohydrate, Premion™, 99.995% (metals basis)

Product Code. 11376967
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376967 1 g 1g
11386967 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376967 Supplier Thermo Scientific Alfa Aesar Supplier No. 010836.03
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 13933-33-0 | Cl2H14N4OPt | 352.12 g/mol

Tetraammineplatinum(II) chloride is used as a catalyst in the chemical synthesis and a precursor for platinum catalysts. This complex has pharmaceutical applications and used in proteomics research. It is used in the preparation of Pt catalysts supported on reduced graphite oxide (RGO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13933-33-0
Molecular Formula Cl2H14N4OPt
Molecular Weight (g/mol) 352.12
MDL Number MFCD00149947
InChI Key GWBDNMCYHWRSOH-UHFFFAOYSA-L
Synonym azane;platinum 2+ ;dichloride;hydrate, tetrammineplatinum ii chloride hydrate, tetraammineplatinum ii chloride monohydrate, platinum 2+ ion tetraamine hydrate dichloride, platinum 2+ tetraamine hydrate dichloride, platinum 2+ chloride-ammonia-water 1/2/4/1, tetraamineplatinium ii chloride
PubChem CID 21946612
IUPAC Name azane;platinum(2+);dichloride;hydrate
SMILES N.N.N.N.O.[Cl-].[Cl-].[Pt++]

Specifications

Melting Point 250°C (dec.)
Color White through Cream
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water
Formula Weight 352.13 (334.11 Anhydrous)
Percent Purity 99.995%
Grade Premion
Sensitivity Hygroscopic
Density 2.74g/mL
Assay (metals basis)
Chemical Name or Material Tetraammineplatinum(II) chloride monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.