missing translation for 'onlineSavingsMsg'
Learn More

Tetraamminepalladium(II) chloride monohydrate, 99.9% (metals basis), Pd 39% min

Product Code. 11341668
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
2 g
10 g
Pakningsstørrelse:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11341668 10 g 10g
11331668 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11341668 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. 011036.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13933-31-8 | Cl2H14N4OPd | 263.46 g/mol

Tetraamminepalladium(II) chloride monohydrate is used to prepare trans-diamminedichloropalladium(II). It is also utilized in the preparation of mesoporous carbon supporting palladium nanoparticles by using graphite oxide. It is used as catalysts, reagents, and analytical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13933-31-8
Molecular Formula Cl2H14N4OPd
Molecular Weight (g/mol) 263.46
MDL Number MFCD00151033
InChI Key WVCXSPJPERKPJS-UHFFFAOYSA-L
IUPAC Name palladium(2+) tetraamine hydrate dichloride
SMILES N.N.N.N.O.[Cl-].[Cl-].[Pd++]

Specifications

Melting Point 120°C (decomposition)
Color Colorless to Yellow
Density 1.91 g/mL
Quantity 10 g
UN Number UN3077
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 263.45 (245.43 Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 39% min.
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tetraamminepalladium(II) chloride monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.