missing translation for 'onlineSavingsMsg'
Learn More

Tetraamminepalladium(II) chloride monohydrate, 99.9% (metals basis), Pd 39% min, Thermo Scientific Chemicals

Product Code. 11331668
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Tetraamminepalladium(II) chloride monohydrate is used to prepare trans-diamminedichloropalladium(II). It is also utilized in the preparation of mesoporous carbon supporting palladium nanoparticles by using graphite oxide. It is used as catalysts, reagents, and analytical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13933-31-8
Molecular Formula Cl2H14N4OPd
Molecular Weight (g/mol) 263.46
MDL Number MFCD00151033
InChI Key WVCXSPJPERKPJS-UHFFFAOYSA-L
IUPAC Name palladium(2+) tetraamine hydrate dichloride
SMILES N.N.N.N.O.[Cl-].[Cl-].[Pd++]

Specifications

Melting Point 120°C (decomposition)
Color Colorless to Yellow
Density 1.91 g/mL
Quantity 2 g
UN Number UN3077
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 263.45 (245.43 Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 39% min.
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tetraamminepalladium(II) chloride monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'