missing translation for 'onlineSavingsMsg'
Learn More

Tetraallylsilane, 97%

Product Code. 11334878
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334878 5 g 5g
11324878 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334878 Supplier Thermo Scientific Alfa Aesar Supplier No. 043183.06
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthesis of dendrimers | CAS: 1112-66-9 | C12H20Si | 192.377 g/mol

It is used in synthesis of dendrimers and as an intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-66-9
Molecular Formula C12H20Si
Molecular Weight (g/mol) 192.377
MDL Number MFCD00048161
InChI Key AKRQMTFHUVDMIL-UHFFFAOYSA-N
Synonym tetraallylsilane, silane, tetra-2-propenyl, tetrakis prop-2-en-1-yl silane, tetrakis prop-2-enyl silane, silane, tetraallyl, tetraallyl silane, zlchem 610, acmc-20ajk5, tetra 2-propenyl silane, silane,tetra-2-propen-1-yl
PubChem CID 565641
IUPAC Name tetrakis(prop-2-enyl)silane
SMILES C=CC[Si](CC=C)(CC=C)CC=C

Specifica

Boiling Point 91°C (10mmHg)
Physical Form Liquid
Quantity 5 g
Flash Point 77°C (171°F)
Refractive Index 1.485
Density 0.8314 g/mL
Solubility Information Insoluble in water.
Formula Weight 192.38
Percent Purity 97%
Chemical Name or Material Tetraallylsilane

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.