missing translation for 'onlineSavingsMsg'
Learn More

Tetraacetylethylenediamine, 90%

Product Code. 10224951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10224951 250 g Plastic bottle 250g
10454955 50 g Plastic bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10224951 Supplier Thermo Scientific Acros Supplier No. 344592500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10543-57-4 | C10H16N2O4 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10543-57-4
Molecular Formula C10H16N2O4
Molecular Weight (g/mol) 228.25
MDL Number MFCD00014967
InChI Key BGRWYDHXPHLNKA-UHFFFAOYSA-N
Synonym tetraacetylethylenediamine, n,n,n',n'-tetraacetylethylenediamine, taed, tetracetylethylenediamine, acetamide, n,n'-1,2-ethanediylbis n-acetyl, n,n'-ethylenebis n-acetylacetamide, unii-p411ed0n2b, n,n'-ethane-1,2-diyl bis n-acetylacetamide, n-acetyl-n-2-diacetylamino ethyl acetamide
PubChem CID 66347
IUPAC Name N-acetyl-N-[2-(diacetylamino)ethyl]acetamide
SMILES CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C

Specifications

Melting Point 149°C to 154°C
Color White to Beige
Boiling Point 140°C (2.0 hPa)
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 88% min
Packaging Plastic bottle
Quantity 250 g
Merck Index 15, 9158
Solubility Information Solubility in water: ca 1g/L (20°C). Other solubilities: slightly soluble in acetone,moderately soluble in chloroform,methylenechloride
Formula Weight 228.25
Percent Purity 90%
Physical Form Granular Powder
Chemical Name or Material Tetraacetylethylenediamine, 90%
Show More Show Less
Is this Tetraacetylethylenediamine (TAED), 90% (Cat. Nos. 344592500, 344590500) uncoated ?

Yes, this Tetraacetylethylenediamine, 90% is uncoated.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.