missing translation for 'onlineSavingsMsg'
Learn More

Tetra-tert-butyldiphenoquinone, TRC

Product Code. 30426040
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426040 50 mg 50mg
30416878 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426040 Supplier Toronto Research Chemicals Supplier No. TRCT28922550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2455-14-3
Molecular Formula C28 H40 O2
Molecular Weight (g/mol) 408.62
Synonym Diphenoquinone, 3,3',5,5'-tetra-tert-butyl- (6CI,7CI), [Bi-2,5-cyclohexadien-1-ylidene]-4,4'-dione, 3,3',5,5'-tetra-tert-butyl- (8CI), 3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone, 3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone, 3,3',5,5'-Tetra-tert-butyldiphenoquinone, 3,5,3',5'-Tetra-tert-butyldiphenoquinone, NSC 14478, Tetra-tert-butyldiphenoquinone
IUPAC Name 2,6-ditert-butyl-4-(3,5-ditert-butyl-4-oxocyclohexa-2,5-dien-1-ylidene)cyclohexa-2,5-dien-1-one
SMILES CC(C)(C)C1=CC(=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)C=C(C1=O)C(C)(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C28H40O2/c1-25(2,3)19-13-17(14-20(23(19)29)26(4,5)6)18-15-21(27(7,8)9)24(30)22(16-18)28(10,11)12/h13-16H,1-12H3
Formula Weight 408.3028
Chemical Name or Material 2,2',6,6'-Tetra-tert-butyldiphenoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.