missing translation for 'onlineSavingsMsg'
Learn More

Tetra-O-(tert-butyldimethyl)silyl cis-Piceatannol, TRC

Product Code. 30409707
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409707 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409707 Supplier Toronto Research Chemicals Supplier No. TRCT29128010MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetra-O-(tert-butyldimethyl)silyl cis-Piceatannol
CAS 106325-84-2
Molecular Formula C38H68O4Si4
Quantity 10 mg
Synonym (Z)-[[4-[2-[3,5-bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]phenyl]ethenyl]-1,2-phenylene]bis(oxy)]bis[(1,1-dimethylethyl)dimethylsilane
InChI Formula InChI=1S/C38H68O4Si4/c1-35(2,3)43(13,14)39-31-25-30(26-32(28-31)40-44(15,16)36(4,5)6)22-21-29-23-24-33(41-45(17,18)37(7,8)9)34(27-29)42-46(19,20)38(10,11)12/h21-28H,1-20H3/b22-21-
SMILES C[Si](C(C)(C)C)(C)OC1=CC(O[Si](C)(C)C(C)(C)C)=CC(/C=C\C2=CC=C(O[Si](C)(C)C(C)(C)C)C(O[Si](C)(C)C(C)(C)C)=C2)=C1
IUPAC Name (Z)-((4-(3,5-bis((tert-butyldimethylsilyl)oxy)styryl)-1,2-phenylene)bis(oxy))bis(tert-butyldimethylsilane)
Molecular Weight (g/mol) 701.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.