missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium perruthenate(VII), 98%

Product Code. 11420328
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420328 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11420328 Supplier Thermo Scientific Alfa Aesar Supplier No. B24511.88
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 114615-82-6 | C12H36NO4Ru | 359.49 g/mol

Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-propylammonium perruthenate a catalytic oxidant used in conjunction with N-methylmorpholine N-oxide for oxidation of primary & secondary alcohols to aldehydes and ketones.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, reducing agents, heat, organic materials, metal powders, heat.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 160°C (decomposition)
CAS 114615-82-6
Quantity 2.5 g
Sensitivity Hygroscopic
Molecular Formula C12H36NO4Ru
Linear Formula (CH3(CH2)2)4NRuO4
MDL Number MFCD00074914
UN Number UN1479
Synonym 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1-
Solubility Information Insoluble aq. soln..
InChI Key HUCLFLGLPCVDMZ-UHFFFAOYSA-N
SMILES O.O.O.O.[Ru].CCC[N+](CCC)(CCC)CCC
IUPAC Name tetrapropylazanium tetrahydrate ruthenium
Molecular Weight (g/mol) 359.49
PubChem CID 127020979
Formula Weight 351.43
Percent Purity 98%
Chemical Name or Material Tetra-n-propylammonium perruthenate(VII)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetra-n-propylammonium perruthenate (VII)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Tetrapropylammoniumperruthenate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.