missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium chloride, 99+%

Product Code. 11333547
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333547 1 g 1g
11323547 5 g 5g
11343547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11333547 Supplier Thermo Scientific Alfa Aesar Supplier No. 019379.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5810-42-4 | C12H28ClN | 221.813 g/mol

Tetra-n-propylammonium chloride is used as phase-transfer type applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5810-42-4
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.813
MDL Number MFCD00038729
InChI Key FBEVECUEMUUFKM-UHFFFAOYSA-M
Synonym tetrapropylammonium chloride, tetrapropyl ammonium chloride, tetra-n-propylammonium chloride, 1-propanaminium, n,n,n-tripropyl-, chloride, 1-propanaminium, n,n,n-tripropyl-, chloride 1:1, n,n,n-tripropylpropan-1-aminium chloride, tetrapropylazanium chloride, acmc-209m2k, ksc491c1h, tetra-n-propylammonium chloride 1g
PubChem CID 79880
IUPAC Name tetrapropylazanium;chloride
SMILES CCC[N+](CCC)(CCC)CCC.[Cl-]

Specifications

Melting Point 240°C to 242°C
Color White
Odor Odorless
Linear Formula (CH3CH2CH2)4NCl
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water, acetone
Formula Weight 221.87
Percent Purity ≥99%
Physical Form Crystalline
Chemical Name or Material Tetra-n-propylammonium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.