missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium bromide, 98%

Product Code. 11454523
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454523 2500 g 2500g
11434523 100 g 100g
11444523 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454523 Supplier Thermo Scientific Alfa Aesar Supplier No. A11522.0E
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1941-30-6 | C12H28BrN | 266.267 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to Strong oxidizing agents. Stable under ordinary conditions (Hygroscopic)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1941-30-6
Molecular Formula C12H28BrN
Molecular Weight (g/mol) 266.267
MDL Number MFCD00011840
InChI Key BGQMOFGZRJUORO-UHFFFAOYSA-M
Synonym tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
PubChem CID 74745
ChEBI CHEBI:55318
IUPAC Name tetrapropylazanium;bromide
SMILES CCC[N+](CCC)(CCC)CCC.[Br-]

Specifications

Melting Point ∼270°C (decomposition)
Quantity 2500 g
Beilstein 3567846
Sensitivity Hygroscopic
Solubility Information It is soluble in water.
Formula Weight 266.27
Percent Purity ≥98%
Chemical Name or Material Tetra-n-propylammonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.