Learn More
Tetra-n-propylammonium bromide, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Tetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to Strong oxidizing agents. Stable under ordinary conditions (Hygroscopic)
Specifications
Specifications
| Melting Point | ∼270°C (decomposition) |
| Quantity | 500 g |
| Beilstein | 3567846 |
| Sensitivity | Hygroscopic |
| Solubility Information | It is soluble in water. |
| Formula Weight | 266.27 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Tetra-n-propylammonium bromide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.