missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-pentylammonium iodide, 98%

Product Code. 11388955
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388955 5 g 5g
11398955 25 g 25g
11308965 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388955 Supplier Thermo Scientific Alfa Aesar Supplier No. L14088.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2498-20-6 | C20H44IN | 425.483 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2498-20-6
Molecular Formula C20H44IN
Molecular Weight (g/mol) 425.483
MDL Number MFCD00041980
InChI Key FBLZDUAOBOMSNZ-UHFFFAOYSA-M
Synonym tetrapentylammonium iodide, tetraamylammonium iodide, tetra-n-pentylammonium iodide, 1-pentanaminium, n,n,n-tripentyl-, iodide 1:1, tetrapentylazanium iodide, tetra-n-amylammonium iodide, tetraamylammoniumjodid, tetraamylammoniumiodide, acmc-209ggb, 1-pentanaminium, n,n,n-tripentyl-, iodide
PubChem CID 17248
IUPAC Name tetrapentylazanium;iodide
SMILES CCCCC[N+](CCCCC)(CCCCC)CCCCC.[I-]

Specifications

Melting Point 135°C to 139°C
Linear Formula [CH3(CH2)4]4NI
Quantity 5 g
Beilstein 3917412
Sensitivity Light Sensitive
Formula Weight 425.49
Percent Purity 98%
Chemical Name or Material Tetra-n-pentylammonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.