missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-octylammonium bromide, 98+%

Product Code. 11412757
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412757 50 g 50g
11402757 10 g 10g
11422757 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412757 Supplier Thermo Scientific Alfa Aesar Supplier No. A14929.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14866-33-2 | C32H68BrN | 546.807 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tetra-n-octylammonium bromide is associated with benzethonium chloride and used as a disinfectant, in surface treatment and preservative in hospitals and industries. It is used as a surfactant involved in the synthesis of blue-luminescent silicon nanoparticles in order to get uniform particle size. It is utilized as a cationic additive in the preparation of membranes using cyclam derivative 1,4,8,11-tetra(n-octyl)-1,4,8,11-tetraazacyclotetradecane as an ionphore and dibutyl phthalate as plasticizers.

Solubility
Soluble in water, dichloromethane, chloroform, acetone and methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14866-33-2
Molecular Formula C32H68BrN
Molecular Weight (g/mol) 546.807
MDL Number MFCD00011863
InChI Key QBVXKDJEZKEASM-UHFFFAOYSA-M
Synonym tetraoctylammonium bromide, tetra-n-octylammonium bromide, tetraoctylazanium bromide, tetraoctyl ammonium bromide, 1-octanaminium, n,n,n-trioctyl-, bromide, unii-52qw29owoc, 52qw29owoc, tetra n-octyl ammonium bromide, acmc-1bvth, tetra-octylammonium bromide
PubChem CID 2734117
IUPAC Name tetraoctylazanium;bromide
SMILES CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC.[Br-]

Specifications

Melting Point 94°C to 99°C
Assay Percent Range 98%
Quantity 50 g
Beilstein 4169176
Solubility Information Soluble in water,dichloromethane,chloroform,acetone and methanol.
Formula Weight 546.81
Percent Purity ≥98%
Chemical Name or Material Tetra-n-octylammonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.