missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium bromide, 98%

Product Code. 11430183
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430183 50 g 50g
11420183 10 g 10g
11440183 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430183 Supplier Thermo Scientific Alfa Aesar Supplier No. A11372.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4328-13-6 | C24H52BrN | 434.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4328-13-6
Molecular Formula C24H52BrN
Molecular Weight (g/mol) 434.59
MDL Number MFCD00011858
InChI Key SYZCZDCAEVUSPM-UHFFFAOYSA-M
Synonym tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
PubChem CID 78026
IUPAC Name tetrahexylazanium;bromide
SMILES [Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Melting Point ∼95°C to 100°C
Quantity 50 g
Beilstein 3637302
Sensitivity Hygroscopic
Formula Weight 434.6
Percent Purity 98%
Chemical Name or Material Tetra-n-hexylammonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.