missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-heptylammonium bromide, 99%

Product Code. p-7022435
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11492238

Brand: Thermo Scientific Alfa Aesar A15218.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4368-51-8 | C28H60BrN | 490.699 g/mol

Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-heptylammonium bromide is used as a mobile phase for the separation of a series of glucosinolates by reversed-phase ion-pair liquid chromatography. It acts as an ion pair reagent, which is used in the extraction of molybdenum complexes into toluene film. Further, it is used in organic synthesis. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4368-51-8
Molecular Formula C28H60BrN
Molecular Weight (g/mol) 490.699
MDL Number MFCD00011861
InChI Key YQIVQBMEBZGFBY-UHFFFAOYSA-M
Synonym tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1-
PubChem CID 78073
IUPAC Name tetraheptylazanium;bromide
SMILES CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-]

Specifications

Melting Point 87°C to 89°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water and methanol.
Formula Weight 490.7
Percent Purity 99%
Chemical Name or Material Tetra-n-heptylammonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.