missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-dodecylammonium iodide, 98%

Product Code. 11388005
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388005 10 g 10g
11378005 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388005 Supplier Thermo Scientific Alfa Aesar Supplier No. L08153.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40797-39-5 | C48H100IN | 818.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40797-39-5
Molecular Formula C48H100IN
Molecular Weight (g/mol) 818.24
MDL Number MFCD00051856
InChI Key OBMLOTDECOTAGU-UHFFFAOYSA-M
Synonym tetradodecylazanium iodide, tetra-n-dodecylammonium iodide, tetradodecylammonium iodide, tetradodecylazanium,iodide
PubChem CID 3084801
SMILES [I-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC

Specifications

Melting Point 114°C to 118°C
Linear Formula [CH3(CH2)11]4NI
Quantity 10 g
Beilstein 5844188
Sensitivity Light sensitive
IUPAC Name tetradodecylazanium;iodide
Formula Weight 818.23
Percent Purity 98%
Chemical Name or Material Tetra-n-dodecylammonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.