missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butyltin, 96%

Product Code. 10413415 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10413415 500 g Glass bottle 500g
10785801 5 g Glass bottle 5g
10041630 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10413415 Supplier Thermo Scientific Acros Supplier No. 137985000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1461-25-2 | C16H36Sn | 347.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-25-2
MDL Number MFCD00009422
InChI Key AFCAKJKUYFLYFK-UHFFFAOYSA-N
Synonym tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
PubChem CID 15098
IUPAC Name tetrabutylstannane
SMILES CCCC[Sn](CCCC)(CCCC)CCCC

Specifications

Melting Point -97°C
Color Colorless to Straw
Density 1.0570g/mL
Boiling Point 145°C (10.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C16H36Sn
Linear Formula [CH3(CH2)3]4Sn
Refractive Index 1.4725 to 1.4745
Quantity 500 g
Specific Gravity 1.057
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 347.14
Viscosity 4 mPa.s (20°C)
Formula Weight 347.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Tetra-n-butyltin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.