missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butyltin, 95%

Product Code. 11327717
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327717 100 g 100g
11317717 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327717 Supplier Thermo Scientific Alfa Aesar Supplier No. 014115.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1461-25-2 | C16H36Sn | 347.174 g/mol

Tetra-n-butyltin is used in hydrogenolysis reactions and as a transition metal catalyst. It is also used stabilizers in the manufacture of polyvinyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-25-2
Molecular Formula C16H36Sn
Molecular Weight (g/mol) 347.174
MDL Number MFCD00009422
InChI Key AFCAKJKUYFLYFK-UHFFFAOYSA-N
Synonym tetrabutyltin, tetra-n-butyltin, stannane, tetrabutyl, tin, tetrabutyl, tetra-n-butylcin, tetra-n-butylcin czech, unii-qj7y5v377v, ccris 6322, tetra-normal-butyltin, wln: 4-sn-4&4&4
PubChem CID 15098
IUPAC Name tetrabutylstannane
SMILES CCCC[Sn](CCCC)(CCCC)CCCC

Specifications

Melting Point -97°C
Density 1.057 g/mL
Boiling Point 144°C to 146°C (10 mmHg)
Flash Point 107°C (225°F)
Odor Mild
Refractive Index 1.474
Quantity 100 g
UN Number UN2788
Solubility Information Insoluble in water. It is soluble in most common organic solvents.
Formula Weight 347.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tetra-n-butyltin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.