missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylthiuram disulfide, 98%

Product Code. 15407428
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407428 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15407428 Supplier Thermo Scientific Alfa Aesar Supplier No. H60917.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1634-02-2 | C18H36N2S4 | 408.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1634-02-2
Molecular Formula C18H36N2S4
MDL Number MFCD00027191
Quantity 100 g
Sensitivity Air Sensitive
Synonym tetrabutylthiuram disulfide, tetra-n-butylthiuram disulfide, tetrabutylthiuram disulphide, thiuram disulfide tetrabutyl, thiuram, tetrabutyl-, disulfide, tetrabutylthioperoxydicarbamic acid, disulfide, bis dibutylthiocarbamoyl, unii-d969886eab, bis dibutylthiocarbamoyl disulfide, thioperoxydicarbonic diamide h2n c s 2s2 , tetrabutyl
InChI Key PGAXJQVAHDTGBB-UHFFFAOYSA-N
SMILES CCCCN(CCCC)C(=S)SSC(=S)N(CCCC)CCCC
IUPAC Name dibutylcarbamothioylsulfanyl N,N-dibutylcarbamodithioate
Molecular Weight (g/mol) 408.74
PubChem CID 15412
Formula Weight 408.75
Percent Purity 98%
Chemical Name or Material Tetra-n-butylthiuram disulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.