missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylphosphonium iodide, 98%

Product Code. 11442818
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442818 25 g 25g
11432818 5 g 5g
11452818 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442818 Supplier Thermo Scientific Alfa Aesar Supplier No. A16792.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3115-66-0 | C16H36IP | 386.34 g/mol

Tetra-n-butylphosphonium iodide is used as phase transfer catalysts, antistatic agents, curing agents for epoxy resins, flame retardants, detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylphosphonium iodide is used as phase transfer catalysts, antistatic agents, curing agents for epoxy resins, flame retardants, detergents.

Solubility
Soluble in water.

Notes
Hygroscopic. Store at 4°C. Store in the dark. Incompatible with oxidizing agents and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3115-66-0
Molecular Formula C16H36IP
Molecular Weight (g/mol) 386.34
MDL Number MFCD00050246
InChI Key CCIYPTIBRAUPLQ-UHFFFAOYSA-M
Synonym tetrabutylphosphonium iodide, tetra-n-butylphosphonium iodide, phosphonium, tetrabutyl-, iodide, tetrabutylphosphanium iodide, acmc-1crr0, wln: 4p4&4&4 &i, phosphonium,tetrabutyl-, iodide 1:1
PubChem CID 201022
IUPAC Name tetrabutylphosphanium;iodide
SMILES [I-].CCCC[P+](CCCC)(CCCC)CCCC

Specifications

Melting Point 98°C to 100°C
Linear Formula [CH3(CH2)3]4PI
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 386.34
Percent Purity 98%
Chemical Name or Material Tetra-n-butylphosphonium iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.