missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylgermanium, 99%

Product Code. 11357788
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357788 25 g 25g
11367788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357788 Supplier Thermo Scientific Alfa Aesar Supplier No. 034119.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1067-42-1 | C16H36Ge | 301.094 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-42-1
Molecular Formula C16H36Ge
Molecular Weight (g/mol) 301.094
MDL Number MFCD00015224
InChI Key HDVLQIDIYKIVRE-UHFFFAOYSA-N
Synonym tetrabutylgermanium, germane, tetrabutyl, tetra-n-butylgermane, tetra-n-butylgermanium, tetrabutyl-german, tetrabuthylgermanium, tetrabutyl germanium
PubChem CID 70599
IUPAC Name tetrabutylgermane
SMILES CCCC[Ge](CCCC)(CCCC)CCCC

Specifications

Density 0.934 g/mL at 20°C
Boiling Point 277°C to 279°C
Flash Point >110°C (230°F)
Refractive Index 1.4571
Quantity 25 g
Formula Weight 301.05
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetra-n-butylgermanium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.