missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium trifluoromethanesulfonate, 99%, ACROS Organics™

Product Code. 10774543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10774543 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10774543 Supplier Thermo Scientific Acros Supplier No. 312550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

CAS 35895-70-6
Melting Point 111°C to 115°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C18H36F3NO2
Linear Formula (CF3SO3)N[(CH2)3CH3]4
MDL Number MFCD00042585
Quantity 5 g
Synonym tetrabutylammonium triflate, tetrabutylammonium trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulfonate, tetrabutylammonium ion triflate, trifluoromethanesulfonic acid tetrabutylammonium salt, acmc-1aglb, tetrakis but-1-yl ammonium triflate, tetrabutylazanium; trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulphonate
InChI Key WTEXQPWIUJQYJQ-UHFFFAOYSA-M
SMILES [O-]C(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium trifluoroacetate
Molecular Weight (g/mol) 355.49
PubChem CID 2733306
Formula Weight 391.54
Percent Purity ≥98.5%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tetra-n-butylammonium trifluoromethanesulfonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.