missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium tribromide, 98%

Product Code. 11347168
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347168 25 g 25g
11337168 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347168 Supplier Thermo Scientific Alfa Aesar Supplier No. 016258.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38932-80-8 | C48H108Br3N3 | 967.125 g/mol

Tetra-n-butylammonium tribromide is used as a phase transfer catalyst in organic synthesis as well as a mild brominating agent. It acts as a solid source of bromine. It is useful for the preparation of vicinal dibromides from alkenes and alkynes and alfa-bromo acetals. Further, it is utilized to generate hydrogen bromide, which is used as an efficient chemoselective reagent for acetalization of carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38932-80-8
Molecular Formula C48H108Br3N3
Molecular Weight (g/mol) 967.125
MDL Number MFCD00012110
InChI Key SFLXUZPXEWWQNH-UHFFFAOYSA-K
Synonym tetrabutylammonium tribromide, tetra-n-butylammonium tribromide, tetrabutylammoniumtribromide, mono tetrabutylammonium tribromide, tetrabutylamoniumtribromide, acmc-1ct1n, tetrabutylamonium tribromide, tetrabutylazanium tribromide, tetrabutylammonium-tribromide, tetra-butylammonium tribromide
PubChem CID 23500184
IUPAC Name tetrabutylazanium;tribromide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-]

Specifications

Melting Point 74°C to 76°C
Color Yellow
Linear Formula [CH3(CH2)3]4NBr3
Quantity 25 g
UN Number UN3261
Beilstein 3746114
Solubility Information Insoluble in water.
Formula Weight 482.18
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Tetra-n-butylammonium tribromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.