Learn More
Tetra-n-butylammonium sulfate, 50% w/w aq. soln.
Description
Tetra-n-butylammonium sulfate is used as a phase transfer catalyst, neutral pH and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.01 |
| Boiling Point | 84°C |
| Odor | Unpleasant |
| Refractive Index | 1.417 |
| Quantity | 100 mL |
| Solubility Information | Fully miscible. |
| Formula Weight | 580.99 |
| Concentration or Composition (by Analyte or Components) | 50% w/w aq. soln. |
| Physical Form | Liquid |
| Chemical Name or Material | Tetra-n-butylammonium sulfate |
Safety and Handling
- Tetra-n-butylammonium sulfate, 50% w/w aqueous solution
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Bis(tetrabutylammonium) sulphate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.