missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium sulfate, 50% w/w aq. soln.

Product Code. 11328878
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328878 2.5 L 2.5 L
11338878 100 mL 100mL
11430421 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328878 Supplier Thermo Scientific Alfa Aesar Supplier No. 041723.K4
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phase transfer catalyst, neutral pH | CAS: 2472-88-0 | C32H72N2O4S | 581.00 g/mol

Tetra-n-butylammonium sulfate is used as a phase transfer catalyst, neutral pH and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2472-88-0
Molecular Formula C32H72N2O4S
Molecular Weight (g/mol) 581.00
MDL Number MFCD00216632
InChI Key ZXUCBXRTRRIBSO-UHFFFAOYSA-L
Synonym bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate
PubChem CID 11433187
IUPAC Name tetrabutylazanium;sulfate
SMILES [O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 1.01
Boiling Point 84°C
Odor Unpleasant
Refractive Index 1.417
Quantity 2.5 L
Solubility Information Fully miscible.
Formula Weight 580.99
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetra-n-butylammonium sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.